missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Osteoactivin/GPNMB Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2330-SP
This item is not returnable.
View return policy
Description
Osteoactivin/GPNMB Monoclonal specifically detects Osteoactivin/GPNMB in Mouse samples. It is validated for Western Blot.
Specifications
| Osteoactivin/GPNMB | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| NP_444340 | |
| GPNMB | |
| Mouse myeloma cell line NS0-derived recombinant mouse Osteoactivin/GPNMB Lys23-Asn502 Accession # NP_444340 | |
| 25 μg | |
| Primary | |
| Detects mouse Osteoactivin/GPNMB in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human Osteoactivin. | |
| Mouse | |
| Purified |
| Western Blot | |
| 297310 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DC-HIL, glycoprotein (transmembrane) nmb, glycoprotein NMB, GPNMB, HGFIN, HGFINglycoprotein nmb-like protein, NMBosteoactivin, Transmembrane glycoprotein HGFIN, transmembrane glycoprotein NMB | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10457 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction