Learn More
Mouse OSCAR Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody has been used in 2 publications
Brand:  R&D Systems MAB1633-SP
 View more versions of this product
Product Code. 18738849
  • £181.00 / 25µg
Description
OSCAR Monoclonal specifically detects OSCAR in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OSCAR
Monoclonal
LYOPH
Western Blot 1 ug/mL
Q8VBT3
OSCAR
Mouse myeloma cell line NS0-derived recombinant mouse OSCAR Asp19-Asn228 Accession # Q8VBT3
25 μg
Primary
Detects mouse OSCAR in direct ELISAs and Western blots. In Western blots, this antibody shows weak cross-reactivity with recombinant human OSCAR.
Mouse
Purified
Western Blot
218311
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
hOSCAR, MGC33613, osteoclast associated receptor OSCAR-S1, osteoclast associated receptor OSCAR-S2, osteoclast associated, immunoglobulin-like receptor, osteoclast-associated immunoglobulin-like receptor, Osteoclast-associated receptor, PIgR-3, PIGR3, Poly-Ig receptor 3, Polymeric immunoglobulin receptor 3
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
126014
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a