missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse OSCAR Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB1633
This item is not returnable.
View return policy
Description
OSCAR Monoclonal specifically detects OSCAR in Mouse samples. It is validated for Western Blot.
Specifications
| OSCAR | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q8VBT3 | |
| OSCAR | |
| Mouse myeloma cell line NS0-derived recombinant mouse OSCAR Asp19-Asn228 Accession # Q8VBT3 | |
| 500 μg | |
| Primary | |
| Detects mouse OSCAR in direct ELISAs and Western blots. In Western blots, this antibody shows weak cross-reactivity with recombinant human OSCAR. | |
| Mouse | |
| Purified |
| Western Blot | |
| 218311 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| hOSCAR, MGC33613, osteoclast associated receptor OSCAR-S1, osteoclast associated receptor OSCAR-S2, osteoclast associated, immunoglobulin-like receptor, osteoclast-associated immunoglobulin-like receptor, Osteoclast-associated receptor, PIgR-3, PIGR3, Poly-Ig receptor 3, Polymeric immunoglobulin receptor 3 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 126014 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction