missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse OCILRP2/CLEC2i Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF3370
This item is not returnable.
View return policy
Description
OCILRP2/CLEC2i Polyclonal specifically detects OCILRP2/CLEC2i in Mouse samples. It is validated for Western Blot.
Specifications
| OCILRP2/CLEC2i | |
| Polyclonal | |
| Biotin | |
| Q9WVF9 | |
| Clec2i | |
| E. coli-derived recombinant mouse OCILRP2/CLEC2i Thr77-Val217 Accession # Q9WVF9 | |
| 50 μg | |
| Primary | |
| Detects mouse OCILRP2/CLEC2i in Western blots. In Western blots, approximately 25% cross-reactivity with recombinant mouse OCIL is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| CLAX, CLEC2i, CLRG, LLT1, OCIL | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 29121 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction