missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse OCILRP2/CLEC2i Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3370-SP
This item is not returnable.
View return policy
Description
OCILRP2/CLEC2i Polyclonal specifically detects OCILRP2/CLEC2i in Mouse samples. It is validated for Western Blot.
Specifications
| OCILRP2/CLEC2i | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLAX, CLEC2i, CLRG, LLT1, OCIL | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 29121 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q9WVF9 | |
| Clec2i | |
| E. coli-derived recombinant mouse OCILRP2/CLEC2i Thr77-Val217 Accession # Q9WVF9 | |
| 25 μg | |
| Primary | |
| Detects mouse OCILRP2/CLEC2i in direct ELISAs and Western blots. In Western blots, approximately 25% cross-reactivity with recombinant mouse OCIL is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction