missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse OCAM/NCAM2 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB778-SP
This item is not returnable.
View return policy
Description
OCAM/NCAM2 Monoclonal specifically detects OCAM/NCAM2 in Mouse samples. It is validated for Western Blot.
Specifications
| OCAM/NCAM2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| NCAM2, N-CAM-2, NCAM-2, NCAM21MGC51008, neural cell adhesion molecule 2, neural cell adhesion molecule 2 precursor10, Rb-8, RNCAM | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4685 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 125101 | |
| Western Blot 1 ug/mL | |
| O35136 | |
| NCAM2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse OCAM/NCAM2 Leu20-Gly700 (predicted) Accession # O35136 | |
| 25 μg | |
| Primary | |
| Detects mouse OCAM/NCAM2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) ALCAM, rhBCAM, rhEpCAM, rhMCAM, recombinant mouse (rm) NCAM-L1, or rmMAdCAM-1 is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction