missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse NPY4R (NP_032945) VersaClone cDNA

Product Code. 18744979
Change view
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Product Code. Quantity unitSize
18744979 10 μg 10µg
1 options
This item is not returnable. View return policy

Product Code. 18744979

Brand: R&D Systems™ RDC2735

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This shuttle vector contains the complete ORF for the gene of interest, along with a Kozak consensus sequence for optimal translation initiation. It is inserted NotI to AscI. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector.

  • Also known as: Y4; Ppyr1; NYYR-D
  • NPY4R is a member of the G protein-coupled receptor 1 family.
  • It is one of five receptors for neuropeptide Y and peptide YY. NPY4R is expressed at high levels in amygdala, corpus callosum, hippocampus and subthalamic nucleus.
  • Neuropeptide Y is one of the most abundant neuropeptides in the mammalian nervous system and exhibits a diverse range of important physiologic activities, including effects on psychomotor activity, food intake, regulation of central endocrine secretion, and potent vasoactive effects on the cardiovascular system.
  • NPY receptors represent novel molecular therapeutic targets in cancer and obesity.
  • The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
  • For reasearch use only. Not for use in humans.
TRUSTED_SUSTAINABILITY

Specifications

Gene mNpy4r
Accession Number NP_032945
Concentration 10 μg at 0.2 μg/μL
Formulation cDNA is provided in 10 mM Tris-Cl, pH 8.5
Insert Type 1141 bp
Gene ID (Entrez) 5540
Content And Storage Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Species Mouse
Quantity 10 μg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.