missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse NOV/CCN3 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems
Additional Details : Weight : 0.00970kg
Description
NOV/CCN3 Monoclonal specifically detects NOV/CCN3 in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).Specifications
NOV/CCN3 | |
Monoclonal | |
LYOPH | |
ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
Q64299 | |
NOV | |
Mouse myeloma cell line NS0-derived recombinant mouse NOV/CCN3 Ser26-Ile354 Accession # Q64299 | |
500 μg | |
Primary | |
Detects mouse NOV/CCN3 in ELISAs. | |
Mouse | |
Purified |
ELISA | |
231226 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CCN family member 3, CCN3, CCN3IGF-binding protein 9, IBP-9, IGFBP-9, IGFBP9novH, Insulin-like growth factor-binding protein 9, nephroblastoma overexpressed gene, Nephroblastoma-overexpressed gene protein homolog, NOVH, protein NOV homolog | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4856 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
For Research Use Only