missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Notch-2 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB5196-SP
This item is not returnable.
View return policy
Description
Notch-2 Monoclonal specifically detects Notch-2 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| Notch-2 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| O35516 | |
| NOTCH2 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Notch-2 Leu26-Val528 (predicted) Accession # O35516 | |
| 25 μg | |
| Primary | |
| Detects mouse Notch-2 in direct ELISAs. In direct ELISAs, 50-100% cross-reactivity with recombinant human (rh) Notch-2 and no cross-reactivity with rhNotch-1, -2, -3, recombinant mouse (rm) Notch-1, rmNotch-3, recombinant rat (rr) Notch2, or rrDLL1 is observed. | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 605724 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| AGS2, hN2, neurogenic locus notch homolog protein 2, Notch (Drosophila) homolog 2, notch 2Notch homolog 2 (Drosophila), Notch homolog 2, Notch2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4853 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction