missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Nogo Receptor/NgR Biotinylated Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems BAF1440
This item is not returnable.
View return policy
Description
Nogo Receptor/NgR Polyclonal specifically detects Nogo Receptor/NgR in Mouse samples. It is validated for Western Blot.
Specifications
| Nogo Receptor/NgR | |
| Polyclonal | |
| Biotin | |
| Q99PI8 | |
| RTN4R | |
| Mouse myeloma cell line NS0-derived recombinant mouse Nogo Receptor/NgR Cys27-Ser447 Accession # Q99PI8 | |
| 50 μg | |
| Primary | |
| Detects mouse Nogo Receptor/NgR in Western blots. In this format, approximately 25% cross-reactivity with recombinant human Nogo Receptor is observed. | |
| Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| NgR, NgR1, Nogo receptor, Nogo-66 receptor, NogoR, NOGORUNQ330/PRO526, reticulon 4 receptor, reticulon-4 receptor, RTN4R | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 65078 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction