missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse Noggin, Animal-Free Recombinant Protein, PeproTech®

Product Code. 17853623
Click to view available options
Quantity:
100 μg
2 x 500 μg
20 μg
500 μg
Unit Size:
100µg
1mg
20µg
500µg
Pack of 2
This item is not returnable. View return policy

Product Code. 17853623

Brand: Gibco™ AF25038500UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

AF-250-38-1MG will be provided as 2 x 500 μg (AF-250-38-500UG). Recombinant Murine Noggin is a 46.4 kDa disulfide-linked homodimer consisting of two 206 amino acid polypeptide chains. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Noggin is involved in numerous developmental processes, such as neural tube fusion and joint formation. The morphogenesis of organs is initiated by a downgrowth from a layer of epithelial stem cells. This process is achieved through the receipt of signals from 1) a WNT protein (WNT3A) to stabilize beta-catenin; and 2) Noggin, which is a bone morphogenetic protein inhibitor. Noggin mutations in unrelated families with proximal symphalangism (SYM1) and multiple synostoses syndrome (SYNS1) have been identified, which have multiple joint fusion as their principal defect.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P97466
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 18121
Molecular Weight (g/mol) 46.4 kDa
Name Mouse Noggin, Animal-Free
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias NOG; Noggin; Noggin precursor; noggin; LOW QUALITY PROTEIN: noggin; SYM1; symphalangism 1 (proximal); SYNS1; SYNS1A
Common Name Noggin
Gene Symbol NOG
Biological Activity Determined by its ability to inhibit 5.0 ng/ml of BMP-4 induced alkaline phosphatase production by ATDC5 chondrogenic cells. The expected ED50 for this effect is 1.0-2.0 ng/ml of Noggin.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MQHYLHIRPA PSDNLPLVDL IEHPDPIFDP KEKDLNETLL RSLLGGHYDP GFMATSPPED RPGGGGGPAG GAEDLAELDQ LLRQRPSGAM PSEIKGLEFS EGLAQGKKQR LSKKLRRKLQ MWLWSQTFCP VLYAWNDLGS RFWPRYVKVG SCFSKRSCSV PEGMVCKPSK SVHLTVLRWR CQRRGGQRCG WIPIQYPIIS ECKCSC
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.