missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse NKG2A/CD159a Antibody, R&D Systems™

Rat Monoclonal Antibody
Marca: R&D Systems MAB6867
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
NKG2A/CD159a/KLRC1 Monoclonal specifically detects NKG2A/CD159a/KLRC1 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Especificaciones
| NKG2A/CD159a/KLRC1 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q9Z202 | |
| KLRC1 | |
| CHO Chinese hamster ovary cell line transfected with mouse NKG2A/CD159a Accession # Q9Z202 | |
| 100 μg | |
| Primary | |
| Stains mouse NKG2A/CD159a transfectants and not irrelevant transfectants in flow cytometry. | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 705829 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD159 antigen-like family member A, CD159a, CD159a antigen, C-lectin type II protein, killer cell lectin-like receptor subfamily C, member 1, Klrc1, MGC13374, natural killer cell lectin, natural killer group protein 2, NK cell receptor A, NKG2, NKG2-1/B activating NK receptor, NKG2-A, NKG2-A/B type II integral membrane protein, NKG2-A/B-activating NK receptor, NKG2-A/NKG2-B type II integral membrane protein, NKG2AMGC59791, NKG2-B | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3821 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido