missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Neuropoietin/NP Antibody, R&D Systems™
Goat Polyclonal Antibody
£393.97
Specifications
Antigen | Neuropoietin/NP |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
Neuropoietin/NP Polyclonal specifically detects Neuropoietin/NP in Mouse samples. It is validated for Western Blot.Specifications
Neuropoietin/NP | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Mouse | |
Cardiotrophin-2, Ctf2, CTF2P, Gm494, NP | |
Ctf2 | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P83714 | |
100289419 | |
E. coli-derived recombinant mouse Neuropoietin/NP Ala23-Ala204 Accession # P83714 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects mouse Neuropoietin/NP in direct ELISAs and Western blots. |
For Research Use Only