missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Mouse Neuregulin-1/NRG1 ELISA Kit (Chemiluminescence)

Product Code. 18686491 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18686491 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18686491 Supplier Novus Biologicals™ Supplier No. NBP268067

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kit (Chemiluminescence)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 15.63 to 1000 pg/mL
Assay Sensitivity 9.38 pg/mL
Product Type ELISA Kit (Chemiluminescence)
Sample Type Serum, Plasma, Biological Fluids
Specificity This kit recognizes Mouse NRG-1 in. samples. No significant cross-reactivity or interference between Mouse NRG-1 and analogues was observed.
For Use With (Application) ELISA
Gene ID (Entrez) 3084
Gene Symbol NRG1
Quantity 1 Kit
Research Discipline Biologically Active Proteins, Chromatin Research, Glia Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Oligodendrocyte Cell Markers, Signal Transduction
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym ARIA, GGF2, GGFglial growth factor, HGLneu differentiation factor, HRG, HRG1, HRGAneuregulin 1 type IV beta 1 A, MST131, MSTP131, NDFheregulin, alpha (45 kD, ERBB2 p185-activator), neuregulin 1, neuregulin 1 type IV beta 3, pro-neuregulin-1, membrane-bound isoform, pro-NRG1, sensory and motor neuron derived factor, SMDF
Target Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.