missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Netrin-1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 7 publications
Brand: R&D Systems AF1109
Additional Details : Weight : 0.00970kg
Description
Netrin-1 Polyclonal specifically detects Netrin-1 in Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction.Specifications
Netrin-1 | |
Polyclonal | |
Unconjugated | |
AAC52971 | |
NTN1 | |
Mouse myeloma cell line NS0-derived recombinant mouse Netrin-1 Val22-Ala603 Accession # AAC52971 | |
100 μg | |
Primary | |
Detects mouse Netrin-1 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant chicken (rch) Netrin-1 and recombinant human Netrin-1 is observed and less than 1% cross-reactivity with recombinant mouse (rm) Netrin-G1a and rmNetrin-4 is observed. | |
Mouse | |
IgG |
Western Blot, Blockade of Receptor-ligand Interaction | |
LYOPH | |
Western Blot 0.1 ug/mL, Blockade of Receptor-ligand Interaction | |
netrin 1, netrin-1, Netrin1, NTN1, NTN1Lnetrin 1, mouse, homolog of | |
Goat | |
Affinity Purified | |
RUO | |
9423 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only