missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Neprilysin DuoSet ELISA

Product Code. 30021468
Click to view available options
Quantity:
1 Kit (for 15 Plates)
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 30021468

Brand: R&D Systems™ DY1126

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Neprilysin DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Mouse Neprilysin DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse Neprilysin/CD10.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Kit (for 15 Plates)
Species Mouse
Conjugate Biotin
Assay Range 93.80 to 6000 pg/mL
For Use With (Application) ELISA
Gene ID (Entrez) 17380
Synonym Atriopeptidase, CALLA, CALLAmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase), CD10, CD10 antigen, CD10), CD10membrane metallo-endopeptidase variant 1, Common acute lymphocytic leukemia antigen, DKFZp686O16152, EC 3.4.24, EC 3.4.24.11, Enkephalinase, EPN, Leu-19, membrane metallo-endopeptidase, membrane metallo-endopeptidase variant 2, MGC126681, MGC126707, MME, NEPmembrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, neprilysin, Neutral endopeptidase, Neutral Endopeptidase 24.11, NKH1, SFE, Skin fibroblast elastase
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.