missing translation for 'onlineSavingsMsg'
Learn More

Mouse Nectin-4 Antibody, R&D Systems™
GREENER_CHOICE

Rat Monoclonal Antibody

£168.00 - £412.00

Specifications

Antigen Nectin-4/PVRL4
Clone 356704
Concentration LYOPH
Dilution Western Blot 1 ug/mL
Applications Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18763959
GREENER_CHOICE
View Documents
R&D Systems
MAB3116-SP
25 μg
£168.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18122341
View Documents
R&D Systems
MAB3116
100 μg
£412.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Nectin-4/PVRL4 Monoclonal specifically detects Nectin-4/PVRL4 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

Nectin-4/PVRL4
LYOPH
Western Blot
Unconjugated
Rat
Mouse
Q8R007
81607
Mouse myeloma cell line NS0-derived recombinant mouse Nectin-4 Tyr28-Ile349 Accession # Q8R007
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects mouse Nectin-4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human Nectin-4, 10% cross-reactivity with recombinant mouse (rm) Nectin-3, and no cross-reactivity with rmNectin-1 or rmNectin-2 is observed.
356704
Western Blot 1 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
EDSS1, Ig superfamily receptor LNIR, LNIR, LNIRnectin 4, nectin-4, Nectin4, poliovirus receptor-related 4, poliovirus receptor-related protein 4, PRR4, PRR4DKFZp686K05193, PVRL4
PVRL4
IgG2a
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Certifikat

For Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.