missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse NCAM-1/CD56 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB7820
This item is not returnable.
View return policy
Description
NCAM-1/CD56 Monoclonal specifically detects NCAM-1/CD56 in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| NCAM-1/CD56 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P13595 | |
| NCAM1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse NCAM-1/CD56 Leu20-Thr711 Accession # P13595 | |
| 100 μg | |
| Primary | |
| Detects mouse NCAM-1/CD56 in direct ELISAs and Western blots. | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 809220 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD56, CD56/ NCAM-1, CD56 antigen, MSK39, NCAM1, N-CAM-1, NCAM-1, NCAMantigen recognized by monoclonal 5.1H11, neural cell adhesion molecule 1, neural cell adhesion molecule, NCAM | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4684 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction