missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse MUP-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6560
This item is not returnable.
View return policy
Description
MUP-1 Monoclonal specifically detects MUP-1 in Mouse samples. It is validated for Immunocytochemistry.
Specifications
| MUP-1 | |
| Monoclonal | |
| LYOPH | |
| Immunocytochemistry 8-25 ug/mL | |
| NP_112465 | |
| Mup1 | |
| Mouse MUP-1 synthetic peptide Accession # NP_112465 | |
| 100 μg | |
| Primary | |
| Detects mouse MUP-1 in direct ELISAs. | |
| Mouse | |
| Purified |
| Immunocytochemistry | |
| 663323 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Ltn-1, Lvtn-1, major urinary protein 1, major urinary protein 10, major urinary protein 7, MUP 1, Mup-1, MUP1, Mup1 major urinary protein 1, Mup10, Mup7, Mup-a, Up-1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 17840 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction