missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Mouse Multi-tissue Tissue MicroArray (Normal)

Product Code. 18635781 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
5 Slides
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18635781 5 Slides Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18635781 Supplier Novus Biologicals™ Supplier No. NBP278097
  • ONLINE EXCLUSIVE PRICE
    £617.00
    £582.75 / Pack of 5
    Save £34.25
    5% Off
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

The panel contains 5 to 8 different tissues on one slide.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 Slides
Species Mouse
For Use With (Application) Immunohistochemistry, Immunohistochemistry-Frozen, In-situ Hybridization
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.