Learn More
Mouse MMR/CD206 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 44 publications
Brand:  R&D Systems AF2535
412.25 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
Product Code. 18051852
  • £499.00 / 100µg
Description
MMR/CD206/Mannose Receptor Polyclonal specifically detects MMR/CD206/Mannose Receptor in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MMR/CD206/Mannose Receptor
Polyclonal
Unconjugated
Q2HZ94
MRC1
Mouse myeloma cell line NS0-derived recombinant mouse MMR/CD206 Leu19-Ala1388 Accession # Q2HZ94
100 μg
Primary
Detects mouse MMR/CD206 in direct ELISAs and Western blots. In direct ELISAs, less than 45% cross-reactivity with recombinant human MMR is observed.
Mouse
IgG
Western Blot, Flow Cytometry, Immunohistochemistry, CyTOF
LYOPH
Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 5-15 ug/mL, CyTOF-ready
CD206, CLEC13D, CLEC13Dmacrophage mannose receptor 1, C-type lectin domain family 13 member D, mannose receptor, C type 1, MMRCD206 antigen, MRC1
Goat
Affinity Purified
RUO
4360
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.