missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Mimecan Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2949-SP
This item is not returnable.
View return policy
Description
Mimecan/OIF Monoclonal specifically detects Mimecan/OIF in Mouse samples. It is validated for Western Blot.
Specifications
| Mimecan/OIF | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp586P2421, KSPG25, mimecan, mimecan proteoglycan, OGN, OIF, OIFOG, osteoglycin, osteoglycin (osteoinductive factor), Osteoinductive factor, SLRR3A, SLRR3Acorneal keratan sulfate proteoglycan | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4969 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot | |
| 329939 | |
| Western Blot 1 ug/mL | |
| Q62000 | |
| OGN | |
| Mouse myeloma cell line NS0-derived recombinant mouse Mimecan Ala20-Phe298 Accession # Q62000 | |
| 25 μg | |
| Primary | |
| Detects mouse Mimecan in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Mimecan is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction