missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Mimecan Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB2949
Additional Details : Weight : 0.00970kg
Description
Mimecan/OIF Monoclonal specifically detects Mimecan/OIF in Mouse samples. It is validated for Western Blot.Specifications
Mimecan/OIF | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
DKFZp586P2421, KSPG25, mimecan, mimecan proteoglycan, OGN, OIF, OIFOG, osteoglycin, osteoglycin (osteoinductive factor), Osteoinductive factor, SLRR3A, SLRR3Acorneal keratan sulfate proteoglycan | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
4969 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Western Blot | |
329939 | |
Western Blot 1 ug/mL | |
Q62000 | |
OGN | |
Mouse myeloma cell line NS0-derived recombinant mouse Mimecan Ala20-Phe298 Accession # Q62000 | |
500 μg | |
Primary | |
Detects mouse Mimecan in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Mimecan is observed. | |
Mouse | |
Purified |
For Research Use Only