missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Midkine Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF7769
This item is not returnable.
View return policy
Description
Midkine Polyclonal specifically detects Midkine in Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| Midkine | |
| Polyclonal | |
| Unconjugated | |
| P12025 | |
| MDK | |
| E. coli-derived recombinant mouse Midkine Gly89-Asp140 Accession # P12025 | |
| 100 μg | |
| Primary | |
| Detects mouse Midkine in direct ELISAs and Western blots. In direct ELISAs, approximately 20% cross-reactivity with recombinant human Midkine is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Amphiregulin-associated protein, ARAP, MDK, MEK, Midgestation and kidney protein, midkine, midkine (neurite growth-promoting factor 2), MK1, MKARAP, NEGF2, NEGF2FLJ27379, Neurite outgrowth-promoting factor 2, Neurite outgrowth-promoting protein | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 4192 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction