missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse MICL/CLEC12A Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2950-100
This item is not returnable.
View return policy
Description
MICL/CLEC12A Monoclonal specifically detects MICL/CLEC12A in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| MICL/CLEC12A | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD371, CLEC12A, CLL1, CLL-1, C-type lectin domain family 12, member A, DCAL-2, MICL | |
| Mouse myeloma cell line NS0-derived recombinant mouse MICL/CLEC12A with an N-terminal 9-His tag Thr67-Arg267 Accession # Q504P2 | |
| 100 μg | |
| Primary | |
| Detects mouse MICL/CLEC12A in direct ELISAs. | |
| Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| 812303 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q504P2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 160364 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO
Spot an opportunity for improvement?Share a Content Correction