missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse MICL/CLEC12A Alexa Fluor 700-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB2950RN-100UG
This item is not returnable.
View return policy
Description
MICL/CLEC12A Monoclonal specifically detects MICL/CLEC12A in Mouse samples. It is validated for Flow Cytometry.
Specifications
| MICL/CLEC12A | |
| Monoclonal | |
| Alexa Fluor 700 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide. | |
| CD371, CLEC12A, CLL1, CLL-1, C-type lectin domain family 12, member A, DCAL-2, MICL | |
| Mouse myeloma cell line NS0-derived recombinant mouse MICL/CLEC12A, Thr67-Arg267, Accession No. Q504P2 | |
| 100 μg | |
| 160364.0 | |
| Mouse | |
| Supernatant |
| Flow Cytometry | |
| 812303R | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q504P2 | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects mouse MICL/CLEC12A in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction