missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Meteorin-like/METRNL Antibody, R&D Systems™

Rat Monoclonal Antibody
£157.00 - £351.00
Specifications
| Antigen | Meteorin-like/METRNL |
|---|---|
| Clone | 829519 |
| Dilution | Immunohistochemistry 8-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18727169
![]() |
R&D Systems
MAB6679-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18719527
![]() |
R&D Systems
MAB6679 |
100 μg |
£351.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Meteorin-like/METRNL Monoclonal specifically detects Meteorin-like/METRNL in Mouse samples. It is validated for Immunohistochemistry.Specifications
| Meteorin-like/METRNL | |
| Immunohistochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Meteorin like, meteorin, glial cell differentiation regulator-like, meteorin-like protein, METRNL, MGC99788 | |
| METRNL | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 829519 | |
| Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q8VE43 | |
| 284207 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Meteorin-like/METRNL Gln46-Glu311 Accession # Q8VE43 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Meteorin-like/METRNL in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title