missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Meteorin-like/METRNL Alexa Fluor 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC66791U-100UG
This item is not returnable.
View return policy
Description
Meteorin-like/METRNL Monoclonal specifically detects Meteorin-like/METRNL in Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Meteorin-like/METRNL | |
| Monoclonal | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Meteorin like, meteorin, glial cell differentiation regulator-like, meteorin-like protein, METRNL, MGC99788 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse METRNL Gln46-Glu311 Accession # Q8VE43 | |
| RUO | |
| 284207 | |
| Mouse | |
| IgG2b |
| Flow Cytometry | |
| 829535 | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q8VE43 | |
| Rat | |
| 100 μg | |
| Primary | |
| Detects mouse METRNL in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction