missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MCP-5 Mouse Instant ELISA™ Kit

Product Code. 15561967
Click to view available options
Quantity:
128 Tests
Unit Size:
128 tests
This item is not returnable. View return policy

Product Code. 15561967

Brand: Invitrogen™ BMS6007INST

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

  • Aluminum pouch(es) with a Microwell Plate coated with Polyclonal Antibody to mouse MCP-5, Biotin-Conjugate (anti-mouse MCP-5 polyclonal antibody), Streptavidin-HRP and Sample Diluent, lyophilized
  • Aluminum pouch(es) with a mouse MCP-5 Standard curve (coloured)
  • Wash Buffer Concentrate 20x (phosphate-buffered saline with 1% Tween 20)
  • Substrate Solution (tetramethyl-benzidine)
  • Sample Diluent (Use when an external predilution of the samples is needed)
  • Stop Solution (1M Phosphoric acid)
  • Adhesive Plate Covers

Instant ELISA Kit

The Mouse Moncyte Chemoattractant Protein 5 (Ms MCP-5) ELISA quantitates Ms MCP-5 in mouse serum, plasma, cell culture supernatants or other body fluids. The assay will exclusively recognize both natural and recombinant Ms MCP-5. Principle of the method The Mouse MCP-5 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the binding of the second (detector) antibody to the target on a different epitope from the capture antibody. An antibody conjugated with enzyme binds the formed sandwich. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Chemotactic factor that attracts eosinophils, monocytes, and lymphocytes but not neutrophils. Potent monocyte active chemokine that signals through CCR2. Involved in allergic inflammation and the host response to pathogens and may play a pivotal role during early stages of allergic lung inflammation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q62401
Assay Range 15.6-1,000 pg/mL
Assay Sensitivity 2.3 pg/mL
Conjugate Biotin
Product Type ELISA Kit
Sample Type Plasma, Serum, Supernatant
Target Species Mouse
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 20293
Gene Symbol MCP-5,Scya12
Interassay CV 10.7%
Intraassay CV 6.7%
Kit Contents Pre-coated 96 well Microplate, Biotinylated Detection Antibody, SAV-HRP, Sample Diluent, lyophilized, Standard curve strips, Wash Buffer Concentrate, Substrate Solution, Stop Solution, Lysis Buffer 10x, Adhesive Plate Covers
Label or Dye HRP
Quantity 128 Tests
Regulatory Status RUO
Sample Volume Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL
Storage Requirements -20°C
Synonym CCL12, Monocyte chemotactic protein 5, SCYA12
Target Mouse
Test Time 20 min.
Total Assay Time 3 hr. 10 min.
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.