Learn More
Invitrogen™ Mouse MCP-1 ELISA Kits
Description
Assay MCP-1 (mouse monocyte chemoattractant protein-1) levels in mouse serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples with our Thermo Scientific™ Mouse MCP-1 ELISA Kits.
Target: Natural and recombinant mouse MCP-1 (monocyte chemoattractant protein-1)
Format: Colorimetric ELISA (TMB substrate) in clear 96-well strip plate
Measurement: Absorbance at 450nm minus absorbance at 550nm
Assay Range: 16 to 1000pg/mL
Sensitivity: < 4pg/mL
Precision: Intra-assay CV < 10%; inter-assay CV < 10%
Sample Types: Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Total Assay Time: 3 hours
Sample Size: 50μL
Specificity: No significant cross-reaction with mouse G-CSF, GM-CSF, IFNγ, IL-1α, IL-1β, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-9, IL-10, IL-18, Eotaxin, MIP-1α, RANTES or TNFα, or human or rat MCP-1
Includes:Complete Kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates. The Reagent Kit (previously called a “Screening Set”) contains matched antibodies, standard and detection reagents but no coated plates.
Specifications
Specifications
| Assay Range | 16 to 1000pg/mL |
| Assay Sensitivity | <4pg/mL |
| Description | Mouse MCP-1 ELISA Kit |
| Immunoassay Kit Format | Colorimetric ELISA (TMB substrate) with Plates |
| Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum |
| Specificity | No significant cross-reaction with mouse G-CSF, GM-CSF, IFNg, IL-1a, IL-1ß, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-9, IL-10, IL-18, Eotaxin, MIP-1a, RANTES or TNFa, or human or rat MCP-1 |
| Sufficient For | 96 microplate wells |
| Kit Contents | TMB Substrate Solution, 12mL, Streptavidin-HRP Concentrate, 75 μL, Stop Solution, 13mL, 30X Wash Buffer, 50mL, Adhesive Plate Covers, 6 ea, Anti-Mouse MCP-1 Precoated 96-well Strip Plate, 1 ea, Biotinylated Antibody Reagent, 12mL, HRP Dilution Buffer, 13mL, Lyophilized Recombinant Mouse MCP-1 Standard, 2 vials, Sample Diluent, 12mL |
| Measurement | Absorbance at 450nm minus absorbance at 550nm |
| Precision | Intra-assay CV <10%; inter-assay CV <10% |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.