missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Matrilin-4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3380-SP
This item is not returnable.
View return policy
Description
Matrilin-4 Polyclonal specifically detects Matrilin-4 in Mouse samples. It is validated for Western Blot.
Specifications
| Matrilin-4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ14417, MATN4, matrilin 4, matrilin-4, Matrilin4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8785 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| O89029 | |
| MATN4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Matrilin-4 Gln22-Lys624 Accession # O89029 | |
| 25 μg | |
| Primary | |
| Detects mouse Matrilin-4 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human (rh) Matrilin-4 is observed and less than 2% cross-reactivity with recombinant mouse Matrilin-2 and rhMatrilin-3 is observed. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction