Learn More
Mouse Matrilin-4 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand:  R&D Systems AF3380
Additional Details : Weight : 0.00970kg
 View more versions of this product
Product Code. 18059661
  • £405.04 / 100µg
Estimated Shipment: 06-06-2024
to see stock.
Description
Description
Matrilin-4 Polyclonal specifically detects Matrilin-4 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
Matrilin-4
Polyclonal
Unconjugated
O89029
MATN4
Mouse myeloma cell line NS0-derived recombinant mouse Matrilin-4 Gln22-Lys624 Accession # O89029
100 μg
Primary
Detects mouse Matrilin-4 in direct ELISAs and Western blots. In direct ELISAs, approximately 15% cross-reactivity with recombinant human (rh) Matrilin-4 is observed and less than 2% cross-reactivity with recombinant mouse Matrilin-2 and rhMatrilin-3 is observed.
Mouse
IgG
Western Blot
LYOPH
Western Blot 0.1 ug/mL
FLJ14417, MATN4, matrilin 4, matrilin-4, Matrilin4
Goat
Affinity Purified
RUO
8785
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.