missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Mast Cell Protease-6/Mcpt6 Antibody, R&D Systems™

Goat Polyclonal Antibody
£168.00 - £451.00
Specifications
| Antigen | Mast Cell Protease-6/Mcpt6 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 3 ug/mL |
| Applications | Western Blot, Immunoprecipitation, Neutralization |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18776797
![]() |
R&D Systems
AF3736-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18012682
|
R&D Systems
AF3736 |
100 μg |
£451.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mast Cell Protease-6/Mcpt6 Polyclonal specifically detects Mast Cell Protease-6/Mcpt6 in Mouse samples. It is validated for Western Blot, Immunoprecipitation, Neutralization.Specifications
| Mast Cell Protease-6/Mcpt6 | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 3 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| 17229 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Mast Cell Protease-6/Mcpt6 Ala22-Ser276 Accession # P21845 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects Mast Cell Protease-6/Mcpt6 in direct ELISAs and Western blots. In direct ELISAs, approximately 35% cross-reactivity with recombinant human (rh) TPS-1 and rhTPS-B2 is observed and less then 10% cross-reactivity with recombinant mouse MCP-7 is observed. |
| LYOPH | |
| Western Blot, Immunoprecipitation, Neutralization | |
| Unconjugated | |
| RUO | |
| P21845 | |
| Tpsb2 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title