Learn More
Mouse MARCO Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems MAB2956-SP
 View more versions of this product
Product Code. 18794117
  • £167.00 / 25µg
Description
MARCO Monoclonal antibody specifically detects MARCO in Mouse samples. It is validated for Flow Cytometry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MARCO
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS and NaCl with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
macrophage receptor with collagenous structureSCARA2Scavenger receptor class A member 2, member 2, SCARA2
Mouse myeloma cell line NS0-derived recombinant mouse MARCO, Gln70-Ser518, Accession # Q60754
25 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
IgG1
Flow Cytometry, CyTOF
579511
Flow Cytometry 0.25 ug/106 cells, CyTOF-ready
Q60754
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
8685
Mouse
Purified