missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse MAdCAM-1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF993
This item is not returnable.
View return policy
Description
MAdCAM-1 Polyclonal specifically detects MAdCAM-1 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| MAdCAM-1 | |
| Polyclonal | |
| Unconjugated | |
| NP_038619 | |
| MADCAM1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse MAdCAM-1 Gln22-Thr365, Predicted Accession # NP_038619 | |
| 100 μg | |
| Primary | |
| Detects mouse MAdCAM-1 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human (rh) MAdCAM-1, recombinant mouse (rm) ICAM-1, rmICAM-2, rmICAM-5, rmVCAM-1, and rhICAM-3. | |
| Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.25 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| hMAdCAM-1, MACAM1, MAdCAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 8174 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction