missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse MAdCAM-1 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB9931
This item is not returnable.
View return policy
Description
MAdCAM-1 Monoclonal specifically detects MAdCAM-1 in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| MAdCAM-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| hMAdCAM-1, MACAM1, MAdCAM1, MAdCAM-1, mucosal addressin cell adhesion molecule 1, mucosal addressin cell adhesion molecule-1, mucosal vascular addressin cell adhesion molecule 1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8174 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 200530 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| NP_038619 | |
| MADCAM1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse MAdCAM-1 Gln22-Thr365 (predicted) Accession # NP_038619 | |
| 500 μg | |
| Primary | |
| Detects human and mouse MAdCAM-1 in ELISAs. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction