missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse M-CSF Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF416
This item is not returnable.
View return policy
Description
M-CSF Polyclonal specifically detects M-CSF in Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
| M-CSF | |
| Polyclonal | |
| Unconjugated | |
| Q3U4F9 | |
| CSF1 | |
| E. coli-derived recombinant mouse M-CSF Lys33-Glu262 Accession # Q3U4F9 | |
| 100 μg | |
| Primary | |
| Detects mouse M-CSF in direct ELISAs and Western blots. In direct ELISAs, approximately 35% cross-reactivity with recombinant human M-CSF and recombinant rat M-CSF is observed, and less than 1% cross-reactivity with recombinant mouse SCF is observed. | |
| Mouse | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.015-0.035 ug/mL | |
| colony stimulating factor 1 (macrophage), CSF1, CSF-1, Lanimostim, macrophage colony stimulating factor, macrophage colony-stimulating factor 1, MCSF, M-CSF, MCSFlanimostim, MGC31930 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1435 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction