missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse LYVE-1 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 27 publications
Brand: R&D Systems MAB2125-100
This item is not returnable.
View return policy
Description
LYVE-1 Monoclonal specifically detects LYVE-1 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF-ready.
Specifications
| LYVE-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| Q8BHC0 | |
| LYVE1 | |
| BaF/3 mouse pro-B cell line transfected with mouse LYVE-1 Ala24-Thr234 Accession # Q8BHC0 | |
| 100 μg | |
| Primary | |
| Detects mouse LYVE-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse CD44 or recombinant human LYVE-1 is observed. | |
| Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Blockade of Receptor-ligand Interaction, Immunocytochemistry, CyTOF | |
| 223322 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cell surface retention sequence binding protein-1, Cell surface retention sequence-binding protein 1, CRSBP1, CRSBP-1, extracellular link domain containing 1, extracellular link domain-containing 1, Extracellular link domain-containing protein 1, HAR, Hyaluronic acid receptor, lymphatic vessel endothelial hyaluronan receptor 1, lymphatic vessel endothelial hyaluronic acid receptor 1, LYVE1, LYVE-1XLKD1, XLKD1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10894 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction