missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Ly-6G (Gr-1) Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 9 publications
Brand: R&D Systems MAB10371-SP
This item is not returnable.
View return policy
Description
Ly-6G/Ly-6C Monoclonal specifically detects Ly-6G/Ly-6C in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.
Specifications
| Ly-6G (Gr-1) | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| GR, Gr-1, Ly-6, Ly6G | |
| Ly-6G transfected EL-4J cell line Accession # P35461 | |
| 25 μg | |
| Detects mouse Ly-6G in Flow Cytometry. | |
| Mouse | |
| Supernatant |
| Flow Cytometry, CyTOF | |
| 1A8 | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P35461 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction