missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems Mouse Luminex™ Discovery Assay

Product Code. 18402450
Change view
Click to view available options
Quantity:
1 Plex
10 Plex
11 Plex
12 Plex
13 Plex
14 Plex
15 Plex
16 Plex
17 Plex
18 Plex
19 Plex
2 Plex
20 Plex
21 Plex
22 Plex
23 Plex
24 Plex
25 Plex
26 Plex
27 Plex
28 Plex
29 Plex
3 Plex
30 Plex
31 Plex
32 Plex
33 Plex
34 Plex
35 Plex
36 Plex
37 Plex
38 Plex
39 Plex
4 Plex
40 Plex
41 Plex
42 Plex
43 Plex
44 Plex
45 Plex
46 Plex
47 Plex
48 Plex
49 Plex
5 Plex
50 Plex
6 Plex
7 Plex
8 Plex
9 Plex
Unit Size:
1 set
Each
50 product options available for selection
Product selection table with 50 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18402450 35 Plex 1 set
18752537 4 Plex Each
30001359 1 Plex Each
30000895 3 Plex Each
18742014 17 Plex 1 set
18721984 14 Plex 1 set
18711984 6 Plex 1 set
18702224 19 Plex 1 set
18792214 12 Plex 1 set
18742174 9 Plex 1 set
18742094 15 Plex 1 set
18742034 24 Plex 1 set
18771874 21 Plex 1 set
18731874 5 Plex 1 set
18782004 31 Plex 1 set
18761874 16 Plex 1 set
18712184 7 Plex 1 set
18701844 32 Plex 1 set
18782394 11 Plex 1 set
18782374 22 Plex 1 set
18742374 18 Plex 1 set
18742444 27 Plex 1 set
18712104 2 Plex 1 set
18762084 23 Plex 1 set
18781944 8 Plex 1 set
18712404 20 Plex 1 set
18762324 13 Plex 1 set
18732134 10 Plex 1 set
18432420 48 Plex 1 set
18432020 40 Plex 1 set
18442650 37 Plex 1 set
18472020 42 Plex 1 set
18411330 45 Plex 1 set
18432220 34 Plex 1 set
18491660 50 Plex 1 set
18471860 30 Plex 1 set
18432110 26 Plex 1 set
18482440 28 Plex 1 set
18451490 49 Plex 1 set
18491200 47 Plex 1 set
18402280 43 Plex 1 set
18482180 46 Plex 1 set
18492200 25 Plex 1 set
18462510 29 Plex 1 set
18481820 38 Plex 1 set
18412080 36 Plex 1 set
18451390 44 Plex 1 set
18472610 41 Plex 1 set
18491820 33 Plex 1 set
18421490 39 Plex 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 50 options available.
50 options
This item is not returnable. View return policy
Product Code. 18402450 Supplier R&D Systems Supplier No. LXSAMSM35

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Luminex bead-based multiplex assays are designed to provide accurate, reproducible results for every target analyte.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 35 Plex
Species Mouse
Binding Target Multiplex Cytokine Assay Kits
Content And Storage Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.