missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse LRP-6 Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB2960
This item is not returnable.
View return policy
Description
LRP-6 Monoclonal specifically detects LRP-6 in Mouse samples. It is validated for Western Blot.
Specifications
| LRP-6 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O88572 | |
| LRP6 | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse LRP-6 Met1-Pro1368 Accession # O88572 | |
| 100 μg | |
| Primary | |
| Detects mouse LRP-6 in direct ELISAs and Western blots. In direct ELISAs, this antibody shows approximately 50 - 60% cross-reactivity with recombinant human (rh) LRP-6 and no cross-reactivity with rhLDL R. | |
| Mouse | |
| Purified |
| Western Blot | |
| 290913 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ADCAD2, FLJ90062, FLJ90421, low density lipoprotein receptor-related protein 6, low-density lipoprotein receptor-related protein 6, LRP6, LRP-6 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4040 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction