missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Mouse LOX-1/OLR1 (NP_619589) VersaClone cDNA
Description
- Also known as: LOX-1; SR-EI; Scare1
- OLR1 is a low density lipoprotein receptor that belongs to the C-type lectin superfamily.
- It is regulated through the cyclic AMP signaling pathway. OLR1 binds, internalizes and degrades oxidized low-density lipoprotein.
- It may be involved in the regulation of Fas-induced apoptosis and may play a role as a scavenger receptor.
- Mutations of OLR1 have been associated with atherosclerosis, risk of myocardial infarction, and may modify the risk of Alzheimer's disease.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | mOlr1 |
| Accession Number | NP_619589 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1105 bp |
| Gene ID (Entrez) | 4973 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Mouse |
| Product Line | R&D Systems |
| Quantity | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?