missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse LOX-1/OLR1 ELISA Kit
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Packungsgröße:
Each
Pack of 10
Pack of 96
Beschreibung
Mouse LOX-1/OLR1 quantitates mouse LOX-1/OLR1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant mouse LOX-1/OLR1.
LOX-1 (lectin-like oxidized LDL receptor -1) is a 40-50 kDa type II membrane glycoprotein class E scavenger receptor expressed by endothelial cells, macrophages, activated platelets and smooth muscle cells. LOX-1 binds to a wide range of ligands, including oxidized low-density lipoprotein (oxLDL), hypochlorite modified high-density lipoprotein (HDL), aged /apoptotic cells, activated platelets and bacteria, reflecting its versatile physiological functions.
Spezifikation
Spezifikation
| Accession Number | CLEC8A; C-type lectin domain family 8 member A; hLOX 1; hLO x 1; hLOX-1; lectin-like oxidized LDL receptor; lectin-like oxidized LDL receptor 1; lectin-like oxidized low-density lipoprotein receptor; Lectin-like oxidized low-density lipoprotein receptor-1; lectin-like oxLDL receptor 1; lectin-type oxidized LDL receptor 1; LOX 1; LO x 1; LOX-1; LOXIN; Oldlr1; Oldr1; Olr1; ox LDL receptor 1; oxidised low density lipoprotein (lectin-like) receptor 1; oxidized low density lipoprotein (lectin-like) receptor 1; oxidized low density lipoprotein receptor 1; oxidized low-density lipoprotein receptor 1; Oxidized low-density lipoprotein receptor 1, soluble form; ox-LDL receptor 1; SCARE1; scavenger receptor class E, member 1; sLOX 1; SLO x 1; soluble LOX 1; SR-EI |
| Assay Range | 0.42 to 10 ng/mL |
| Assay Sensitivity | 0.045 ng/mL |
| Conjugate | HRP |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 108078 |
| Gene Symbol | OLR1 |
| Interassay CV | <12% |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction