missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse LOX-1/OLR1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB15641-SP
This item is not returnable.
View return policy
Description
LOX-1/OLR1 Monoclonal specifically detects LOX-1/OLR1 in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| LOX-1/OLR1 | |
| Monoclonal | |
| LYOPH | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| AAG44998 | |
| OLR1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse LOX-1/OLR1 Arg60-Ile363 Accession # AAG44998 | |
| 25 μg | |
| Primary | |
| Detects mouse LOX-1/OLR1 in ELISAs. In sandwich immunoassays, no cross-reactivity or interference with recombinant human LOX-1 or human LDL, HDL or VLDL is observed. | |
| Mouse | |
| Purified |
| ELISA | |
| 214001 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLEC8A, CLEC8ASLOX1, C-type lectin domain family 8 member A, hLOX-1, Lectin-like oxidized LDL receptor 1, Lectin-like oxLDL receptor 1, Lectin-type oxidized LDL receptor 1, LOX1, LOX-1, LOX1ox LDL receptor 1, LOXIN, OLR1, oxidised low density lipoprotein (lectin-like) receptor 1, oxidized low density lipoprotein (lectin-like) receptor 1, oxidized low-density lipoprotein receptor 1, oxidized low-density lipoprotein receptor 1, soluble form, Ox-LDL receptor 1, SCARE1, scavenger receptor class E, member 1, SR-E1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4973 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction