missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Mouse Limitin/IFN-zeta DuoSet ELISA

Product Code. 30021674
Change view
Click to view available options
Quantity:
15 x 96-well Plates
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30021674 15 x 96-well Plates 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30021674 Supplier R&D Systems™ Supplier No. DY597

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sufficient for at least 15 96-well plates

The Mouse Limitin/IFN-zeta DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies mouse Limitin/IFN-zeta.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 15 x 96-well Plates
Conjugate Biotin
Assay Range 15.60 to 1000 pg/mL
For Use With (Application) ELISA
Gene ID (Entrez) 319146
Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.