missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse LIGHT/TNFSF14 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
£371.48
Specifications
Antigen | LIGHT/TNFSF14 |
---|---|
Clone | 261639 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
LIGHT/TNFSF14 Monoclonal specifically detects LIGHT/TNFSF14 in Mouse samples. It is validated for Western Blot.Specifications
LIGHT/TNFSF14 | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Mouse | |
Q9QYH9 | |
8740 | |
Mouse myeloma cell line NS0-derived recombinant mouse LIGHT Asp72-Val239 Accession # Q9QYH9 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse LIGHT in direct ELISAs and Western blots. In Western blots, this antibody shows less than 10% cross-reactivity with recombinant mouse (rm) EDA, rmTRAIL, rmTRANCE, rmTWEAK, and rhVEGI but does not cross-react rm4-1BB Ligand, rhAPRIL, rmBAFF, rhEDA-A2, rmFas Ligand, rhGITR Ligand, rmGITR Ligand, rhLymphotoxin alpha 1 beta 2 rhLymphotoxin alpha 2 beta 2 rmOX40 Ligand, and rmTNF-alpha . |
261639 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CD258, CD258 antigen, delta transmembrane LIGHT, Herpes virus entry mediator ligand, Herpesvirus entry mediator ligand, HVEM-L, HVEMLherpesvirus entry mediator-ligand, ligand for herpesvirus entry mediator, LIGHTherpesvirus entry mediator A, LTg, TNFSF14, TR2, tumor necrosis factor (ligand) superfamily, member 14, tumor necrosis factor ligand superfamily member 14, tumor necrosis factor receptor-like 2, tumor necrosis factor superfamily member LIGHT | |
TNFSF14 | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only