missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Mouse LIGHT, Animal-Free Recombinant Protein, PeproTech®

Product Code. 17893423
Click to view available options
Quantity:
1 mg
100 μg
20 μg
500 μg
Unit Size:
100µg
1mg
20µg
500µg
This item is not returnable. View return policy

Product Code. 17893423

Brand: Gibco™ AF31512100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Murine LIGHT is a 20.1 kDa protein containing 183 amino acid residues, comprising the TNF-homologous region of the LIGHT extracellular domain. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Apoptosis, or programmed cell death, occurs during normal cellular differentiation and development of multicellular organisms. Apoptosis is induced by certain cytokines including TNF (tumor necrosis factor) and Fas ligand of the TNF family through their death domain containing receptors, TNFR1 (tumor necrosis factor receptor 1) and Fas. Cell death signals are transduced by death domain (DD)-containing adapter molecules and members of the ICE (interleukin-1-beta converting enzyme)/CED-3 (C. elegans death receptor-3) protease family. A novel DD-containing molecule was recently cloned from mouse, human and monkey and designated Daxx (death associated protein 6). Daxx binds specifically to the Fas death domain and enhances Fas induced apoptosis and activates the Jun N-terminal kinase (JNK) pathway. Daxx is widely expressed in fetal and adult human and mouse tissues indicating its important function in Fas signaling pathways.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9QYH9
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 50930
Molecular Weight (g/mol) 20.1 kDa
Name Mouse LIGHT, Animal-Free
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias CD258; Herpes virus entry mediator ligand; herpesvirus entry mediator ligand; HVEML; HVEM-L; Light; LTg; Ly113; sCD258; sLIGHT; soluble CD258; soluble LIGHT; Tnfsf14; TR2; tumor necrosis factor (ligand) superfamily, member 14; tumor necrosis factor ligand 1 D; tumor necrosis factor ligand superfamily member 14; Tumor necrosis factor ligand superfamily member 14, membrane form; Tumor necrosis factor ligand superfamily member 14, soluble form; tumor necrosis factor superfamily member 14; UNQ391/PRO726
Common Name LIGHT
Gene Symbol TNFSF14
Conjugate Unconjugated
Recombinant Recombinant
Sequence MRLHQRLGDI VAHLPDGGKG SWEKLIQDQR SHQANPAAHL TGANASLIGI GGPLLWETRL GLAFLRGLTY HDGALVTMEP GYYYVYSKVQ LSGVGCPQGL ANGLPITHGL YKRTSRYPKE LELLVSRRSP CGRANSSRVW WDSSFLGGVV HLEAGEEVVV RVPGNRLVRP RDGTRSYFGA FMV
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.