missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Lgr5/GPR49 Antibody, R&D Systems™

Rat Monoclonal Antibody
£168.00 - £420.00
Specifications
| Antigen | Lgr5/GPR49 |
|---|---|
| Clone | 889901 |
| Dilution | Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready |
| Applications | Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18764058
![]() |
R&D Systems
MAB82401-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18729377
![]() |
R&D Systems
MAB82401 |
100 μg |
£420.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lgr5/GPR49 Monoclonal specifically detects Lgr5/GPR49 in Mouse samples. It is validated for Flow Cytometry, CyTOF-ready.Specifications
| Lgr5/GPR49 | |
| Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FEX, G protein-coupled receptor 49, GPR49, GPR49G-protein coupled receptor HG38, GPR67, G-protein coupled receptor 49, G-protein coupled receptor 67, GRP49, HG38, leucine-rich repeat containing G protein-coupled receptor 5, leucine-rich repeat-containing G protein-coupled receptor 5, leucine-rich repeat-containing G-protein coupled receptor 5, MGC117008, orphan G protein-coupled receptor HG38 | |
| LGR5 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 889901 | |
| Flow Cytometry, CyTOF | |
| Unconjugated | |
| Rat | |
| Mouse | |
| Q9Z1P4 | |
| 8549 | |
| Mouse Lgr5/GPR49 N-terminal peptide Ala21-Cys34 Accession # Q9Z1P4 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Lgr5/GPR49 in direct ELISAs. Stains mouse Lgr5 transfected cells but not irrelevant transfectants in Flow Cytometry. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title