missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Leptin R Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB497
This item is not returnable.
View return policy
Description
Leptin R Monoclonal specifically detects Leptin R in Mouse samples. It is validated for Flow Cytometry, ELISA Capture (Matched Antibody Pair).
Specifications
| Leptin R | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B219, CD295, CD295 antigen, DB, DKFZp686B1731, huB219, LEPR, LEP-R, leptin receptor, LeptinR, OB R, OB receptor, OB-R, OBRCD295 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3953 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Flow Cytometry, ELISA | |
| 55305 | |
| Flow Cytometry 0.25 ug/10^6 cells, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P48356 | |
| LEPR | |
| Mouse myeloma cell line NS0-derived recombinant mouse Leptin R Leu21-Leu840 (predicted) Accession # P48356 | |
| 500 μg | |
| Primary | |
| Detects mouse Leptin R in ELISAs. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction