Learn More
Mouse Legumain/Asparaginyl Endopeptidase Antibody, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody
Brand:  R&D Systems MAB2058-SP
Product Code. 18724329
  • £147.00 / 25µg
Description
Legumain/Asparaginyl Endopeptidase Monoclonal specifically detects Legumain/Asparaginyl Endopeptidase in Mouse samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Legumain/Asparaginyl Endopeptidase
Monoclonal
LYOPH
Western Blot 2 ug/mL
O89017
LGMN
Mouse myeloma cell line NS0-derived recombinant mouse Legumain/Asparaginyl Endopeptidase Val18-Tyr435 Accession # O89017
25 μg
Primary
Detects mouse Legumain/Asparaginyl Endopeptidase in direct ELISAs and Western blots. In Western blots, approximately 25% cross-reactivity with recombinant human Legumain is observed.
Mouse
Purified
Western Blot
301417
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
AEP, Asparaginyl endopeptidase, cysteine protease 1, legumain, LGMN, LGMN1, Protease, cysteine 1, protease, cysteine, 1 (legumain), PRSC1EC 3.4.22.34
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
5641
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2a